missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacillus pumilus ZapA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117636
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Activator of cell division through the inhibition of FtsZ GTPase activity, therefore promoting FtsZ assembly into bundles of protofilaments necessary for the formation of the division Z ring. It is recruited early at mid-cell but it is not essential for cell division.
Specifications
| Bacillus pumilus ZapA | |
| Polyclonal | |
| Unconjugated | |
| zapA | |
| zapA | |
| Recombinant Bacillus pumilus Cell division protein ZapA protein (1-85aa). | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A8FG14 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 58146727 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction