Learn More
BINDER™ Back Access Port
Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Description
- For use in back
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
| Description | Access port with silicone plug - 10 mm, back |
| For Use With (Equipment) | BINDER Chambers FDL 115 |
| Material | Steel |
| Type | Access Port |
| Includes | Silicone plug |
| Diameter (Metric) Inside | 10 mm |