Learn More
BINDER™ Back Access Port
Product Code. 16454159 Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 16454159
Brand: BINDER™ 80121284
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For Use With: FDL 115
  • For use in back
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 10 mm, back
For Use With (Equipment) BINDER Chambers FDL 115
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 10 mm