Learn More
Honeywell™ BACOU TXL REPORTER S3 Shoes
Honeywell™ BACOU TXL REPORTER S3 Shoes feature water-resistant oiled full grain leather uppers and an ergonomic collar.
Brand:  Honeywell™ 6244416-43
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11887030
  • € 101.00 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • 200 J Freelium™ non metallic toe cap
  • Non magnetic
  • Chemically neutral
  • Ultra light: 20% lighter than a standard composite toe-cap
  • Ultra thin: 30% thinner than a standard composite toe-cap
  • Inalterable Excellent thermal insulator Imperceptible to wearers
  • Flexium™ anti-puncture midsole
  • Non-metallic, made with high tenacity textile fibers
  • Covers 100% of the plantar surface
  • The midsole is light and flexible
  • No constraints for the wearer when flexing the foot
  • Antistatic and comfortable
  • Insole is ergonomic and antistatic
  • Anti-bacteria treatment
  • Provides optimal comfort and hygiene
Specifications
Specifications
ISO 9001 / 2008, EC PPE Category 2, EC Attestation Number
Leather
Polyurethane
Black
43
Punctures, static and bacteria