Learn More
Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (05), FITC, Invitrogen™
Mouse Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552323
 View more versions of this product
Product Code. 30019626
  • € 174.00 / Pack of 25
Description
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bacteriophage M13
Recombinant Monoclonal
0.1 mg/mL
PBS with no preservative
Mouse
Protein A
RUO
Virus
Antibody
IgG1
Flow Cytometry
05
FITC
Enterobacteria phage M13
M13 Bacteriophage.
25 Tests
Primary
4°C, store in dark, DO NOT FREEZE!