missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (PS00-01)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544937
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
Specifications
| Bacteriophage M13 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA | |
| PS00-01 | |
| Unconjugated | |
| Enterobacteria phage M13 | |
| M13 Bacteriophage. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction