Learn More
Gibco™ Bacto™ TC Yeastolate
Description
Bacto TC Yeastolate is an animal origin-free hydrolysate from the water-soluble portion of autolyzed yeast. This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins. Derived from the same yeast strain as Difco TC Yeastolate, ultra-filtered (UF), this product has a slightly different nutritional profile because it was not ultra-filtered. Bacto TC Yeastolate should be considered a suitable alternative to Difco TC Yeastolate, ultra-filtered (UF), for supplementing a variety of CHO-based applications.
Customers have used this supplement for CHO-based production of biotherapeutic monoclonal antibodies used in oncological and rheumatological therapeutics and recombinant proteins. Additionally, customers have used this supplement in combination with a soy supplement, such as Difco Phytone, ultra-filtered (UF), to provide an optimal titer performance in mammalian cell based processes by providing additional vitamins and carbohydrates.
Specifications
Specifications
| Content And Storage | Store at 2°C to 25°C in cool, dry conditions in well-sealed containers. |
| Cell Type | Bacteria |
| Product Type | TC Yeastolate |
| Product Line | Bacto |
| Quantity | 100 g |
| Source | Animal origin-free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.