missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baculovirus Envelope gp64 protein Antibody (AcV1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00208APC
This item is not returnable.
View return policy
Description
Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.
Specifications
| Baculovirus Envelope gp64 protein | |
| Monoclonal | |
| APC | |
| ACNVgp129, AcOrf-128 | |
| The immunogen for this antibody is baculovirus envelope gp64 protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| AcV1 | |
| Flow Cytometry | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction