missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAF57 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2591JF646
This item is not returnable.
View return policy
Description
BAF57 Polyclonal antibody specifically detects BAF57 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BAF57 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BAF57BRG1-associated factor 57, chromatin remodeling complex BRG1-associated factor 57, subfamily e, member 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily E member 1 | |
| The immunogen recognized by this antibody maps to a region between residue 360 and the C-terminus (residue 411) of human BAF57/SMARCE1using the numbering given in entry NP_003070.3 (GeneID 6605). | |
| 0.1 mL | |
| Apoptosis, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 6605 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction