missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (2403C) [DyLight 405], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB1162E
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| BAFFR/TNFRSF13C | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 2403C | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C | |
| Mouse myeloma cell line, NS0-derived human BAFF R/TNFRSF13C, Ser7-Ala71, Accession # Q96RJ3 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
| 115650 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction