missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (7J21), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22005APCCY7
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry.
Specifications
| BAFFR/TNFRSF13C | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein G purified | |
| Primary | |
| Mouse | |
| IgG2 |
| Flow Cytometry | |
| 7J21 | |
| Flow Cytometry | |
| TNFRSF13C | |
| Mouse recombinant BAFFR/TNFRSF13C | |
| 0.1 mL | |
| 115650 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction