missing translation for 'onlineSavingsMsg'
Learn More

BAFFR/TNFRSF13C Antibody (BAFFR/1558), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08868AF350

Product Code. 18513316

  • ONLINE EXCLUSIVE PRICE
    € 643.00
    € 607.95 / 100µL
    Save € 35.05
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAFFR/TNFRSF13C
Monoclonal
Alexa Fluor 350
BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C
Recombinant full-length human BAFFR/TNFRSF13C protein (Uniprot: Q96RJ3)
100 μg
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
115650
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, Immunohistochemistry (Paraffin)
BAFFR/1558
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.