missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bag-1 Antibody (SR1542), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Bag-1 |
|---|---|
| Clone | SR1542 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631955
|
Novus Biologicals
NBP3-22527-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661434
|
Novus Biologicals
NBP3-22527-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bag-1 Monoclonal antibody specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Bag-1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| A synthesized peptide derived from human Bag-1 (Uniprot #: Q99933) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1542 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 573 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title