missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG5 Monoclonal Antibody (OTI3B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810633S
This item is not returnable.
View return policy
Description
BAG5 Monoclonal antibody specifically detects BAG5 in Human samples. It is validated for Western BlotSpecifications
| BAG5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UL15 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3B10 | |
| Unconjugated | |
| BAG5 | |
| 1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein | |
| Full length human recombinant protein of human BAG5 produced in 293T. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?