missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG5 Monoclonal Antibody (OTI3B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810633
This item is not returnable.
View return policy
Description
It has been hypothesizes that the BAG-5 protein will induce the death of nigral neurons through its predicted interaction with hsp70, which will cause increased protein aggregation and cell death by disinhibition of hsp70′s anti-apoptotic function. It is believed that BAG-5 will play an important role in the mechanisms of neuronal death. BAG-5 may also be of interest due to its possible role as a modulator of the hsp70/hsp40 chaperone axis or its possible interaction and coordination of localization/ modulation of other BAG containing proteins via BAG-BAG heterodimerization.Specifications
| BAG5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UL15 | |
| BAG5 | |
| Full length human recombit protein of human BAG5 produced in 293T. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3B10 | |
| Unconjugated | |
| BAG5 | |
| BAG-5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction