missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAG6 Polyclonal Antibody

Product Code. 16578871
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16578871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16578871 Supplier Invitrogen™ Supplier No. PA596033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MEPNDSTSTA VEEPDSLEVL VKTLDSQTRT FIVGAQMNVK EFKEHIAASV SIPSEKQRLI YQGRVLQDDK KLQEYNVGGK VIHLVERAPP QTHLPSGASS GTGSASATHG GGSPPGTRGP GASVHDRNAN SYVMVGTFNL PSDGSAVDVH INMEQAPIQS EPRVRLVMAQ HMIRDIQTLL SRMECRGGPQ PQHSQPPPQP; Positive Samples: 293T, HeLa, Mouse testis, Rat testis; Cellular Location: Cytoplasm, Nucleus, cytosol.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Bag6
Gene Accession No. P46379, Q6MG49, Q9Z1R2
Gene Alias 2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe
Gene Symbols Bag6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human BAT3/BAT3/BAG6 (NP_0010920041).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224727, 7917, 94342
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.