Learn More
Invitrogen™ BAG6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114180
Description
Immunogen sequence: VGDPPQPLPE EPMEVQGAER ASPEPQRENA SPAPGTTAEE AMSRGPPPAP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 92%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 93%; Pig: 92%; Rabbit: 93%; Rat: 93%.
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| BAG6 | |
| Polyclonal | |
| Unconjugated | |
| Bag6 | |
| 2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7917 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P46379 | |
| Bag6 | |
| Immunogen sequence: VGDPPQPLPE EPMEVQGAER ASPEPQRENA SPAPGTTAEE AMSRGPPPAP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.