missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Antibody (480912), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB49691APCCY7
This item is not returnable.
View return policy
Description
BAI1 Monoclonal antibody specifically detects BAI1 in Human samples. It is validated for Flow Cytometry
Specifications
| BAI1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 480912 | |
| Flow Cytometry | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| HEK293 human embryonic kidney cell line transfected with human BAI1, Ala31-Thr879, Accession # O14514 | |
| 0.1 mL | |
| Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience | |
| 575 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction