missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI2 Antibody (2528B) [DyLight 405], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9338E
This item is not returnable.
View return policy
Description
BAI2 Monoclonal antibody specifically detects BAI2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| BAI2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 2528B | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2 | |
| Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3 | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR, Neuroscience | |
| 576 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction