missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI2 Antibody (2528B) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9338JF669
This item is not returnable.
View return policy
Description
BAI2 Monoclonal antibody specifically detects BAI2 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| BAI2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2 | |
| Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 2528B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR, Neuroscience | |
| 576 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction