missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI3 Antibody (409611), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3965Z
This item is not returnable.
View return policy
Description
BAI3 Monoclonal antibody specifically detects BAI3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| BAI3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 409611 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
| Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242 | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR | |
| 577 | |
| Store at 4°C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido