missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI3 Antibody (409633), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB39651W
This item is not returnable.
View return policy
Description
BAI3 Monoclonal antibody specifically detects BAI3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| BAI3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 409633 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
| Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242 | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR | |
| 577 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction