missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAIAP3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41080CL1
This item is not returnable.
View return policy
Description
BAIAP3 Polyclonal antibody specifically detects BAIAP3 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| BAIAP3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| BAI1-associated protein 3, BAI-associated protein 3, BAP3KIAA0734MGC138334 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human BAP3. The immunogen is located within amino acids 60 - 110 of BAP3. Amino Acid Squence: CRQSPHPGTHTQV | |
| 0.1 mL | |
| Neuroscience | |
| 8938 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction