missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAK Antibody (8D1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26482-50ul
This item is not returnable.
View return policy
Description
BAK Monoclonal antibody specifically detects BAK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Specifications
| BAK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 8D1 | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| Apoptosis regulator BAK, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
| A synthesized peptide derived from Human BAK [UniProt Q16611] | |
| 50 μL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 578 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction