Learn More
Bode Chemie™ Baktolan™ Cleansing Lotion
Product Code. 11336261
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Product Type:
Cleansing Lotion
Sensitive Cleansing Lotion
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Product Type unitSize
11336261 100 mL Cleansing Lotion 100mL
15179786 500 mL Sensitive Cleansing Lotion 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11336261 Supplier Bode Chemie™ Supplier No. 9811373
  • ONLINE EXCLUSIVE PRICE
    € 15.95
    € 7.91 / 100mL
    Save € 8.04
    50% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Keep skin smooth and pliable even with intensive and repeated daily use with Bode Chemie™ Baktolan™ Cleansing Lotion with Allantoin and wheat protein. The pH value of 5.5 preserves the sensitive protective acid mantle of the skin.
  • Water-in-oil-in-water emulsion
  • Protects against aqueous solutions and regenerates cracked, heavily stressed skin
  • Innovative recipe (W / O / W emulsion)
  • Protection against water-soluble substances
  • Support of skin regeneration
  • Strengthening the skin barrier
  • Very good skin tolerance
  • Free of perfume, silicone and dyes
  • Counteracts occlusion-related irritation under the glove
Specifications
Product Type Cleansing Lotion
Packaging Tube
pH Range Approximately 5.5
Quantity 1 Ea.
Volume (Metric) 100 mL