Learn More
Bode Chemie™ Baktolin™ Hand Wash Lotion
Product Code. 15151436
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15151436 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15151436 Supplier Bode Chemie™ Supplier No. 9813282
Specifications
Form Lotion
pH ∼5.5
Composition Aqua, sodium Laureth sulfate, sodium chloride, PEG-7 Glyceryl Cocoate, Cocamidopropyl betain, Glycer
Container Type Bottle
Volume (Metric) 500 mL