missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAL/PARP9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92245-0.1ml
This item is not returnable.
View return policy
Description
BAL/PARP9 Polyclonal antibody specifically detects BAL/PARP9 in Human samples. It is validated for Western Blot
Specifications
| BAL/PARP9 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| PARP-9, poly (ADP-ribose) polymerase family, member 9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human PARP9 (NP_001139577.1). MEIKKETAAEILFDEVLTFAKDHVKHQLTVKFVIFPTDLEIYKAFSSEMAKRSKMLSLNNYSVPQSTREEKRENGLEARSPAINLMGFNVEEMYEAHAWIQRILSLQNHHIIENNHILYLGRKEHDILSQLQKTSSVSITEIISPGRTELEIEGARADLIEVVMNIEDMLCKVQEEMARKKERGLWRSLGQWTIQQQKTQDEMKENIIFLKCPVPPTQELLDQKKQFEKCGLQVLKVEKIDNEVLMAAFQRKKKMMEEKLHRQPVSHRLFQQVPYQFCNVVCRVGFQRMYSTPCDPKYGAG | |
| 0.1 mL | |
| Apoptosis | |
| 83666 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction