missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAMBI Monoclonal Antibody (OTI9G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807760
This item is not returnable.
View return policy
Description
BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.Specifications
| BAMBI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13145 | |
| BAMBI | |
| Human recombit protein fragment corresponding to amino acids 21-152 of human BAMBI produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9G4 | |
| Unconjugated | |
| BAMBI | |
| NMA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25805 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction