missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Band 3 Antibody (Q1/156), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08610V
This item is not returnable.
View return policy
Description
Band 3 Monoclonal antibody specifically detects Band 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Band 3 | |
| Monoclonal | |
| DyLight 405 | |
| AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group | |
| Human fetal liver cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Q1/156 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6521 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction