missing translation for 'onlineSavingsMsg'
Learn More

Band 3 Antibody (Q1/156), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50113F

Product Code. 18191859

  • ONLINE EXCLUSIVE PRICE
    € 466.00
    € 439.95 / 0.10mL
    Save € 26.05
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Band 3 Monoclonal specifically detects Band 3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Band 3
Monoclonal
FITC
PBS with 0.05% Sodium Azide
SLC4A1
Foetal liver cells
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Q1/156
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Mouse
Protein A purified
RUO
6521
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.