missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BANP Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA521030
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A suggested positive control is mouse kidney tissue lysate. PA5-21030 can be used with blocking peptide PEP-1144.
This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifica
| BANP | |
| Polyclonal | |
| Unconjugated | |
| BANP | |
| AA408158; Banp; BEN domain-containing protein 1; BEND1; BTG3 associated nuclear protein; Btg3-associated nuclear protein; Protein BANP; scaffold/matrix-associated region-1-binding protein; Smar1; SMARBP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292064, 53325, 54971 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8N9N5, Q8VBU8 | |
| BANP | |
| A 14 amino acid peptide near the center of human BANP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto