missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP1 Antibody (1G8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60521AF532
This item is not returnable.
View return policy
Description
BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| BAP1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BAP1 | |
| Full-length human BAP1 protein expressed and purified from E.coli. [UniProtNo. Q92560] | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| 1G8 | |
| Western Blot, Flow Cytometry | |
| BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
| Mouse | |
| 0.1 mL | |
| 8314 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction