missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | BAP1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BAP1/2667 |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4), Tween-20, 0.2% BSA |
| Gene Alias | BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 |
| Host Species | Mouse |
| Immunogen | Recombinant human BAP1 protein fragment (aa191-aa326, exact sequence is proprietary) |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?