missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82671-25ul
This item is not returnable.
View return policy
Description
BAP18 Polyclonal specifically detects BAP18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BAP18 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| BAP18, BPTF-associated protein of 18 kDa, chromatin complexes subunit BAP18, chromosome 17 open reading frame 49, MGC49942, MLL1/MLL complex subunit C17orf49 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C17ORF49 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TSASTKVGEIFSAAGAAFTKLGELTMQLHPVADSSPAGAKWTETEIEMLRAAVKRFGDDLNHISCVIKERTVAQIKATVKRKVYEDSGI | |
| 25 μL | |
| DNA replication Transcription Translation and Splicing | |
| 124944 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction