missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human UBE2I/Ubc9 His (N-Term) Protein

Product Code. 18492222 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18492222

Brand: Novus Biologicals™ NBP23509410ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE
Formulation A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 7.6, with 125 mM NaCl, 10 % Glycerol, 1 mM DTT.
Gene ID (Entrez) 7329
Molecular Weight (g/mol) 19.5 kDa
Quantity 10 μg
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of UBE2I/Ubc9 as determined by LAL method.
Gene Alias C358B7.1, EC 6.3.2, EC 6.3.2.-, P18, SUMO-1-protein ligase, SUMO-protein ligase, UBC9SUMO-conjugating enzyme UBC9, UBCE9, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, ubiquitin conjugating enzyme 9, Ubiquitin-conjugating enzyme E2 I, ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9), ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast), ubiquitin-conjugating enzyme UbcE2A, ubiquitin-like protein SUMO-1 conjugating enzyme, ubiquitin-protein ligase E2I, Ubiquitin-protein ligase I
Gene Symbol UBE2I
Reconstitution Please contact technical support for detailed reconstitution instructions.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein UBE2I/Ubc9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.