missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAP29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562029
This item is not returnable.
View return policy
Description
Immunogen sequence: KLVEDQEKLK TELRKTSDAL SKAQNDVMEM KMQSERLSKE YDQLLKEHSE LQDRLERGNK K Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 82%.
Bap29 and the related protein Bap31 are endoplasmic reticulum (ER) and ER-vesicle membrane proteins and members of the B-cell receptor-associated protein family. These two proteins are highly homologous and can form homo- and heterodimers. Both Bap29 and Bap31 interact with membrane-bound immunoglobulins (mIgs), such as IgM and IgD, which with Ig-alpha/Ig-beta heterodimers form B cell antigen receptors. Binding of the Bap29/Bap31 heterodimer correlates with the ER retention of non-Ig-alpha/Ig-beta bound mIg complexes, suggesting that Bap29 and Bap31 may act as chaperones transmembrane regions of various proteins. Bap29 possesses multiple isoforms.
Specifications
| BAP29 | |
| Polyclonal | |
| Unconjugated | |
| Bcap29 | |
| AW208404; B cell receptor associated protein 29; BAP29; Bcap29; B-cell receptor-associated protein 29; B-cell receptor-associated protein BAP29; BCR-associated protein 29 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHQ4 | |
| Bcap29 | |
| Recombinant protein corresponding to Human BAP29. Recombinant protein control fragment (Product #RP-103052). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction