missing translation for 'onlineSavingsMsg'
Learn More

Barbadin, TRC

Product Code. 30135499
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135499 50 mg 50mg
30008284 2.5 mg 2.5mg
30005152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135499 Supplier Toronto Research Chemicals Supplier No. TRCB11825050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356568-70-2
Molecular Formula C19H15N3OS
Molecular Weight (g/mol) 333.41
Synonym 3-Amino-5-[4-(phenylmethyl)phenyl]thieno[2,3-d]pyrimidin-4(3H)-one
IUPAC Name 3-amino-5-(4-benzylphenyl)thieno[2,3-d]pyrimidin-4-one
SMILES O=C(N(C=N1)N)C2=C1SC=C2C3=CC=C(CC4=CC=CC=C4)C=C3

Specifications

Analyte or Component Names Barbadin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C19H15N3OS/c20-22-12-21-18-17(19(22)23)16(11-24-18)15-8-6-14(7-9-15)10-13-4-2-1-3-5-13/h1-9,11-12H,10,20H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 333.0936
Percent Purity >95%
Chemical Name or Material Barbadin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.