Learn More
Masterflex™ Y-Connector, Barbed, Polypropylene
Click to view available options
Color:
Clear
White
Packaging Quantity:
10/Pk.
20/Pk.
35 mm
Tubing Size:
1/16 in. ID
1/8 in. ID
10mm ID
16mm ID
3/32 in. ID
4-6mm ID
4mm ID
6mm ID
Unit Size:
Pack of 10
Pack of 20
Description
Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.
- Ideal for the following:
- Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
- Designed for quick and easy assembly with flexible tubing
- Available in a variety of barb sizes, fitting lengths, and fitting widths
Specifications
Specifications
| Overall Length (Metric) | 32 mm |
| For Use With | 4-6mm ID Tubing |
| For Use With (Application) | Low pressure applications. Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing |
| Packaging Quantity | 10/Pk. |
| Product Line | Tubing connector |
| Tubing Size | 4-6mm ID |
| Hose Barb | 6mm, 6mm, 6mm |
| Sterilization Method | Autoclave |
| Width (Metric) | 36 mm |
| Shape | Y |
| Show More |