Learn More
Saint Gobain Life Sciences BarbLock™ Hand Tool
Click to view available options
Product Line:
125 Series
187 Series
250 Series
252 Series
500 Series
Unit Size:
Each
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
Specifications
Specifications
| Product Line | 250 Series |
| Product Type | Tubing Clamp |