missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BARD1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-319AF594
This item is not returnable.
View return policy
Description
BARD1 Polyclonal antibody specifically detects BARD1 in Human samples. It is validated for Western Blot
Specifications
| BARD1 | |
| Polyclonal | |
| Western Blot | |
| BARD-1, BRCA1 associated RING domain 1, BRCA1-associated RING domain gene 1, BRCA1-associated RING domain protein 1, EC 6.3.2.- | |
| The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human BRCA1-associated RING Domain 1 using the numbering given in entry NP_000456.1 (GeneID 580). | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 580 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction