missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Barecetamab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

€ 477.00 - € 1204.00

Specifications

Antigen Barecetamab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283064
GREENER_CHOICE
View Documents
Invitrogen™
MA559413
100 μg
€ 477.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283054
View Documents
Invitrogen™
MA559414
1 mg
€ 1204.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Barecetamab, also known as ISU-104, is a fully human anti-ErbB3 monoclonal antibody. The ERBB3 gene, located at 12q13 on chromosome 12, encodes a transmembrane tyrosine kinase receptor that is part of the ERBB receptor family. The receptor produced by ERBB3 has a molecular weight of approximately 180 kDa and features a ligand-binding extracellular domain (ECD) with four subdomains, a transmembrane region, and an intracellular domain. Unlike other ERBB family receptors, ERBB3 lacks intrinsic kinase activity, but it can still activate downstream signaling pathways by forming heterodimers with other ErbB receptors such as EGFR and HER2. This activation is pivotal in various cellular processes, including cell proliferation and differentiation. The gene is of particular interest in oncology, as aberrant expression and mutations have been implicated in cancer pathogenesis and treatment resistance.
Specifications

Specifications

Barecetamab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado