missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, ACS Reagent Grade, Scharlab

Product Code. 11339863
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339863 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11339863 Supplier Scharlab Supplier No. BA00550500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry for the determination of sulfates.

  • ACS Reagent Grade
  • Molecular Formula: BaCl2 • 2H2O
  • Molar Mass: 244.28g/mol
  • CAS: 10326-27-9
  • EIN ECS No.: 233-788-1
  • Oxidizing substances: 0.005% max

Specifications

pH 5.0–8.2 (5%, water)
CAS 10326-27-9
Melting Point 962°C (release of crystalline water)
Quantity 500 g
Molecular Formula BaCl2H4O2
MDL Number MFCD00149154
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
Solubility Information In water: 357g/L at 20°C
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES O.O.[Cl-].[Cl-].[Ba++]
IUPAC Name barium(2+) dihydrate dichloride
Molecular Weight (g/mol) 244.26
PubChem CID 5284346
ChEBI CHEBI:86153
Formula Weight 244.28g/mol
Percent Purity ≥99% (complexometric)
Grade ACS Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Barium chloride dihydrate, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302+H332-Harmful if swallowed or if inhaled
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.