Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
Brand:  Thermo Scientific™ 25001-DB
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 14885701
  • € 103.00 / Each
Estimated Shipment: 26-08-2024
to see stock.
Add to basket
Description
Description
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
Specifications
Specifications
Barnstead Classic Stills
Ventguard Filter Element
Ventguard Filter Element
Fits Ventgard Air Filter