Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Product Code. 11796259 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
E-Pure 3-holder
E-Pure 3-holder, Organic free
E-Pure 4-holder
E-Pure 4-holder, Organic free
E-Pure Ultrapure Water Purification System
Feedwater Source:
N/A
Pretreated
Pretreated feed
Tap
Unit Size:
Each
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Feedwater Source unitSize
11796259 E-Pure 4-holder, Organic free Tap Each
12341278 E-Pure 3-holder, Organic free Pretreated feed Each
11476348 E-Pure 4-holder Pretreated Each
10277244 E-Pure 4-holder, Organic free Pretreated Each
11786259 E-Pure 4-holder Tap Each
10086614 E-Pure 3-holder Pretreated feed Each
11796289 E-Pure Ultrapure Water Purification System N/A Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11796259 Supplier Thermo Scientific™ Supplier No. D5023
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
TRUSTED_SUSTAINABILITY
Specifications
Description Cartridge kit for E-Pure 4-holder, Tap feed, Organic free
Feedwater Source Tap
For Use With (Equipment) E-Pure 4-holder, Organic free
Type Cartridge Kit