Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Product Code. p-4531451 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
E-Pure 3-holder
E-Pure 3-holder, Organic free
E-Pure 4-holder
E-Pure 4-holder, Organic free
E-Pure Ultrapure Water Purification System
Feedwater Source:
N/A
Pretreated
Pretreated feed
Tap
Unit Size:
Each
Pack of 3
Product Code. 12341278
Brand: Thermo Scientific™ D5022
Only null left
Add to basket
Add to basket
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
TRUSTED_SUSTAINABILITY
Specifications
Description Cartridge kit for E-Pure 3-holder, Pretreated feed, Organic
Feedwater Source Pretreated feed
For Use With (Equipment) E-Pure 3-holder, Organic free
Type Cartridge Kit