Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Shop All Thermo Scientific Products
Click to view available options
Type:
Calibration Module
Deionized Water Feed (Low Organics, Type I Water)
Deionized Water Feed (Ultralow Organics, Type I Water)
Low Water Level Pump Protector
R/O and Distilled Feed (Low Organics, Type I Water)
R/O and Distilled Feed (Ultralow Organics, Type I Water)
Unit Size:
Each
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
Specifications
| For Use With (Equipment) | Nanopure Water Purifications System |
| Type | R/O and Distilled Feed (Low Organics, Type I Water) |