Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilters
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
€ 446.00
Specifications
Type Ultrafilter
View More Specs
Products 1
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11317171
View Documents Promotion Details
Thermo Scientific™
50133981
MicroPure, Smart2Pure (UF and UF/UV units only)
€ 446.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
Specifications
Specifications
Ultrafilter