missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basal Bodies of Cillia Antibody (LhS 28), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50114AF750
This item is not returnable.
View return policy
Description
Basal Bodies of Cillia Monoclonal specifically detects Basal Bodies of Cillia in Human, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Basal Bodies of Cillia | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BHK a21 Cytoskeletal preparation | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| LhS 28 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Neuroscience | |
| Human, Hamster | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction