Learn More
Basal Bodies of Cillia Antibody (LhS 28) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50114
 View more versions of this product
Product Code. 18639646
  • € 351.00 / 0.10mL
Description
Basal Bodies of Cillia Monoclonal antibody specifically detects Basal Bodies of Cillia in Human, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Basal Bodies of Cillia
Monoclonal
1 mg/mL
Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Immunofluorescence, Immunoprecipitation
LhS 28
Unconjugated
PBS
BHK a21 Cytoskeletal preparation
0.1 mL
Neuroscience
Human, Hamster