Description
Basalin Polyclonal specifically detects Basalin in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | Basalin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Gene Alias | basalin, intermediate filament-associated protein, Protein S100-A17, S100 calcium binding protein A17, S100 calcium-binding protein A17, S100A17, THHL1, trichohyalin-like 1, trichohyalin-like protein 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Basalin fragment. |
| Gene ID (Entrez) | 126637 |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Isotype | IgG |
| Show More |