Description
Bottom or base for attaching racks for various vessels, such as test tubes, micro reaction vessels, etc., to the platform of the selected platform shaker. Tool-free assembly.
Material: electro-polished stainless steel
Characteristics: autoclavable and dishwasher safe
Specifications
Specifications
| Product Type | Tube Holder Base |
| Material | Stainless Steel |
| For Use With (Equipment) | Hei-SHAKE 1000 or 2000 Series |